2-Chloro-4-formylpyridine ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Chloropyridine-4-carboxaldehyde, 2-Chloroisonicotinaldehyde
C2723-1G
C2723-5G
TCIAC2723-1GEA
915
NOK
TCIAC2723-1G
TCIAC2723-5G
2-Chloro-4-formylpyridine ≥98.0% (by GC)
2-Chloro-4-formylpyridine
Formel:
C₆H₄ClNO MW: 141,56 g/mol |
MDL nummer:
MFCD06651557 CAS-nummer: 101066-61-9 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 47.0 to 51.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...