4-Chloropyridine-2-carbonitrile ≥98.0% (by GC)

Leverandør: TCI
Danger

C2732-1G C2732-5G
TCIAC2732-1GEA 715 NOK
TCIAC2732-1G TCIAC2732-5G
4-Chloropyridine-2-carbonitrile ≥98.0% (by GC)
4-Chloropyridine-2-carbonitrile
Formel: C₆H₃ClN₂
MW: 138,5500 g/mol
MDL nummer: MFCD06009833
CAS-nummer: 19235-89-3
FN: 3439
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 82.0 to 86.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR