(S)-(+)-2-Chloromandelic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
(S)-2-(2-Chlorophenyl)-2-hydroxyacetic acid
C2738-5G
C2738-25G
TCIAC2738-5GEA
1575
NOK
TCIAC2738-5G
TCIAC2738-25G
(S)-(+)-2-Chloromandelic acid ≥98.0% (by GC, titration analysis)
(S)-(+)-2-Chloromandelic acid
Formel:
C₈H₇ClO₃ MW: 186,59 g/mol |
MDL nummer:
MFCD00798437 CAS-nummer: 52950-19-3 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 119.0 to 123.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...