4-Chloro-3-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
C2760-1G
C2760-5G
TCIAC2760-1GEA
435
NOK
TCIAC2760-1G
TCIAC2760-5G
4-Chloro-3-(trifluoromethyl)benzaldehyde ≥97.0% (by GC)
4-Chloro-3-(trifluoromethyl)benzaldehyde
Formel:
C₈H₄ClF₃O MW: 208,57 g/mol Kokepunkt: 54°C |
MDL nummer:
MFCD00040722 CAS-nummer: 34328-46-6 EINECS: 251-940-5 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...