4-Chloro-N-methylpicolinamide ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4-Chloro-N-methylpyridine-2-carboxamide
C2793-1G
C2793-5G
TCIAC2793-1GEA
1275
NOK
TCIAC2793-1G
TCIAC2793-5G
4-Chloro-N-methylpicolinamide ≥98.0% (by HPLC, titration analysis)
4-Chloro-N-methylpicolinamide
Formel:
C₇H₇ClN₂O MW: 170,60 g/mol |
MDL nummer:
MFCD02185921 CAS-nummer: 220000-87-3 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...