(2-Carboxyethyl)phenylphosphinic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 3-(Hydroxy(phenyl)phosphoryl)propanoic acid, 3-Hydroxyphenylphosphinylpropanoic acid

C2887-5G C2887-25G
TCIAC2887-5GEA 890 NOK
TCIAC2887-5G TCIAC2887-25G
(2-Carboxyethyl)phenylphosphinic acid ≥98.0% (by HPLC, titration analysis)
(2-Carboxyethyl)phenylphosphinic acid
Formel: C₉H₁₁O₄P
MW: 214,16 g/mol
MDL nummer: MFCD08704562
CAS-nummer: 14657-64-8
EINECS: 411-200-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR