4-Cyano-4'-ethylbiphenyl ≥98.0% (by GC)

Leverandør: TCI
Warning

C2892-5G C2892-25G
TCIAC2892-5GEA 755 NOK
TCIAC2892-5G TCIAC2892-25G
4-Cyano-4'-ethylbiphenyl ≥98.0% (by GC)
4-Cyano-4'-ethylbiphenyl
Formel: C₁₅H₁₃N
Kokepunkt: 205°C
MDL nummer: MFCD00799420
CAS-nummer: 58743-75-2
EINECS: 261-414-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR