4-Cyano-4'-propoxybiphenyl ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4'-Propoxy-[1,1'-biphenyl]-4-carbonitrile

C2911-1G C2911-5G
TCIAC2911-1GEA 340 NOK
TCIAC2911-1G TCIAC2911-5G
4-Cyano-4'-propoxybiphenyl ≥98.0% (by GC)
4-Cyano-4'-propoxybiphenyl
Formel: C₁₆H₁₅NO
MW: 237,3 g/mol
MDL nummer: MFCD00183206
CAS-nummer: 52709-86-1
EINECS: 258-122-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR