6-Chloro-1-tetralone ≥96.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
6-Chloro-3,4-dihydronaphthalen-1(2H)-one, 6-Chloro-3,4-dihydro-1(2H)-naphthalenone
C2917-1G
C2917-5G
TCIAC2917-1GEA
580
NOK
TCIAC2917-1G
TCIAC2917-5G
6-Chloro-1-tetralone ≥96.0% (by GC)
6-Chloro-1-tetralone
Formel:
C₁₀H₉ClO MW: 180,63 g/mol Kokepunkt: 150°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD04114377 CAS-nummer: 26673-31-4 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Orange powder to lump |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...