4-Chloro-6-fluoroquinoline ≥98.0% (by GC)

Leverandør: TCI
Warning

C2989-1G C2989-5G
TCIAC2989-1GEA 795 NOK
TCIAC2989-1G TCIAC2989-5G
4-Chloro-6-fluoroquinoline ≥98.0% (by GC)
4-Chloro-6-fluoroquinoline
Formel: C₉H₅ClFN
MW: 181,60 g/mol
MDL nummer: MFCD00278783
CAS-nummer: 391-77-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR