Carteolol hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 5-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-2(1H)-quinolinone hydrochloride

C3015-50MG C3015-250MG
TCIAC3015-50MGEA 1100 NOK
TCIAC3015-50MG TCIAC3015-250MG
Carteolol hydrochloride ≥98.0% (by HPLC, titration analysis)
Carteolol hydrochloride
Formel: C₁₆H₂₄N₂O₃·HCl
MW: 328,84 g/mol
Smeltepunkt: 278°C
MDL nummer: MFCD00941499
CAS-nummer: 51781-21-6
EINECS: 257-415-7
Merck Indeks: 14,01866

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR