Cloperastine hydrochloride ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-Chlorobenzhydryl 2-(1-Piperidyl)ethyl ether hydrochloride, 1-(2-((4-Chlorophenyl)(phenyl)methoxy)ethyl)piperidine hydrochloride, 1-[2-[(4-Chlorophenyl)(phenyl)methoxy]ethyl]piperidine hydrochloride

C3038-5G C3038-25G
TCIAC3038-5GEA 555 NOK
TCIAC3038-5G TCIAC3038-25G
Cloperastine hydrochloride ≥97.0% (by HPLC, titration analysis)
Cloperastine hydrochloride
Formel: C₂₀H₂₄ClNO·HCl
MW: 366,33 g/mol
MDL nummer: MFCD00079012
CAS-nummer: 14984-68-0
EINECS: 239-067-8
Merck Indeks: 14,02395

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR