4-Chloro-2-methylbenzyl alcohol ≥98.0% (by GC)

Leverandør: TCI
Warning

C3069-1G C3069-5G
TCIAC3069-1GEA 580 NOK
TCIAC3069-1G TCIAC3069-5G
4-Chloro-2-methylbenzyl alcohol ≥98.0% (by GC)
4-Chloro-2-methylbenzyl alcohol
Formel: C₈H₉ClO
MW: 156,61 g/mol
MDL nummer: MFCD06201140
CAS-nummer: 129716-11-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 64.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR