2-Chloro-N-methylaniline ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
C3121-1G C3121-5G
TCIAC3121-1GEA 405 NOK
TCIAC3121-1G TCIAC3121-5G
2-Chloro-N-methylaniline ≥98.0% (by GC, titration analysis)
2-Chloro-N-methylaniline
Formel: C₇H₈ClN
MW: 141,6 g/mol
Kokepunkt: 218°C
Flash punkt: 92°C
MDL nummer: MFCD00045170
CAS-nummer: 932-32-1
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %