2-Cyano-5-pyridineboronic acid pinacol ester ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 2-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile, 2-(6-Cyano-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-Cyano-3-pyridylboronic acid Pinacol ester, 2-Cyanopyridine-5-boronic acid pinacol ester

C3126-200MG C3126-1G
TCIAC3126-200MGEA 460 NOK
TCIAC3126-200MG TCIAC3126-1G
2-Cyano-5-pyridineboronic acid pinacol ester ≥98.0% (by titrimetric analysis)
2-Cyano-5-pyridineboronic acid pinacol ester
Formel: C₁₂H₁₅BN₂O₂
MW: 230,07 g/mol
MDL nummer: MFCD06657825
CAS-nummer: 741709-63-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80aac2a96738d0e3a83855cb43f5fae23594f94c&vid=80aac2a96738d0e3a83855cb43f5fae23594f94c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 110.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR