2-Chloro-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
2-(6-Chloro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Chloropyridine-5-boronic acid Pinacol ester, 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
C3136-1G
C3136-5G
TCIAC3136-1GEA
515
NOK
TCIAC3136-1G
TCIAC3136-5G
2-Chloro-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Chloro-5-pyridineboronic acid pinacol ester
Formel:
C₁₁H₁₅BClNO₂ MW: 239,51 g/mol |
MDL nummer:
MFCD03412793 CAS-nummer: 444120-94-9 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 85.0 to 89.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...