4-Cyano-4'-ethoxybiphenyl ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
4'-Ethoxy-[1,1'-biphenyl]-4-carbonitrile
C3153-5G
C3153-25G
TCIAC3153-5GEA
1025
NOK
TCIAC3153-5G
TCIAC3153-25G
4-Cyano-4'-ethoxybiphenyl ≥98.0% (by GC)
4-Cyano-4'-ethoxybiphenyl
Formel:
C₁₅H₁₃NO MW: 223,2700 g/mol |
MDL nummer:
MFCD01218033 CAS-nummer: 58743-78-5 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Mesomorphic range | 98.0 to 106.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...