6,8-Dichloro-3-cyanochromone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 6,8-Dichloro-3-cyanochromone, 6,8-Dichloro-4-oxo-4H-1-benzopyran-3-carbonitrile, 6,8-Dichlorochromone-3-carbonitrile

C3178-1G C3178-5G
TCIAC3178-1GEA 475 NOK
TCIAC3178-1G TCIAC3178-5G
6,8-Dichloro-3-cyanochromone ≥98.0% (by GC)
6,8-Dichloro-3-cyanochromone
Formel: C₁₀H₃Cl₂NO₂
MDL nummer: MFCD00192004
CAS-nummer: 72798-32-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 176.0 to 180.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR