4-Carboxy-2-fluorophenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 4-Carboxy-2-fluorobenzeneboronic acid

C3220-1G C3220-5G
TCIAC3220-1GEA 785 NOK
TCIAC3220-1G TCIAC3220-5G
4-Carboxy-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
4-Carboxy-2-fluorophenylboronic acid
Formel: C₇H₆BFO₄
MW: 183,93 g/mol
MDL nummer: MFCD08458473
CAS-nummer: 851335-07-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a194abd5ab130fabe02e00cd5b7bd59693f4eef2&vid=a194abd5ab130fabe02e00cd5b7bd59693f4eef2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 97.0 to 111.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR