5-Cyano-2-fluorophenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 5-Cyano-2-fluorobenzeneboronic acid

C3227-1G C3227-5G
TCIAC3227-1GEA 1200 NOK
TCIAC3227-1G TCIAC3227-5G
5-Cyano-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
5-Cyano-2-fluorophenylboronic acid
Formel: C₇H₅BFNO₂
MW: 164,9300 g/mol
MDL nummer: MFCD03095147
CAS-nummer: 468718-30-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f4d497dbe50ee7198528ab9ee1ea554798a443f&vid=7f4d497dbe50ee7198528ab9ee1ea554798a443f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR