(Chloromethyl)dimethoxy(methyl)silane ≥97.0% (by GC)

Leverandør: TCI
Warning

C3256-1G C3256-5G
TCIAC3256-1GEA 220 NOK
TCIAC3256-1G TCIAC3256-5G
(Chloromethyl)dimethoxy(methyl)silane ≥97.0% (by GC)
(Chloromethyl)dimethoxy(methyl)silane
Formel: C₄H₁₁ClO₂Si
MW: 154,67 g/mol
Kokepunkt: 140°C
Flash punkt: 55°C
MDL nummer: MFCD04974117
CAS-nummer: 2212-11-5
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38ebfdb6ca93d7284f35616854fb4ce0e5c82815&vid=38ebfdb6ca93d7284f35616854fb4ce0e5c82815&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR