5-Chloro-2-hydroxy-4-methylbenzophenone ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
C3260-5G
C3260-25G
TCIAC3260-5GEA
595
NOK
TCIAC3260-5G
TCIAC3260-25G
5-Chloro-2-hydroxy-4-methylbenzophenone ≥98.0% (by GC)
5-Chloro-2-hydroxy-4-methylbenzophenone
Formel:
C₁₄H₁₁ClO₂ |
MDL nummer:
MFCD00002325 CAS-nummer: 68751-90-6 EINECS: 272-140-2 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 140.0 to 144.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...