2-Chloro-4-fluoro-5-nitrotoluene ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
C3273-5G
C3273-25G
TCIAC3273-5GEA
620
NOK
TCIAC3273-5G
TCIAC3273-25G
2-Chloro-4-fluoro-5-nitrotoluene ≥98.0% (by GC)
2-Chloro-4-fluoro-5-nitrotoluene
Formel:
C₇H₅ClFNO₂ |
MDL nummer:
MFCD11110549 CAS-nummer: 112108-73-3 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 38.0 to 42.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...