2-Chloro-3-fluoro-5-methylpyridine ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
C3275-1G
C3275-5G
TCIAC3275-1GEA
365
NOK
TCIAC3275-1G
TCIAC3275-5G
2-Chloro-3-fluoro-5-methylpyridine ≥98.0% (by GC)
2-Chloro-3-fluoro-5-methylpyridine
Formel:
C₆H₅ClFN MW: 145,56 g/mol Kokepunkt: 92°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD06658238 CAS-nummer: 34552-15-3 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...