2-Chloro-4-iodobenzoic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

C3281-1G C3281-5G
TCIAC3281-1GEA 540 NOK
TCIAC3281-1G TCIAC3281-5G
2-Chloro-4-iodobenzoic acid ≥98.0% (by titrimetric analysis)
2-Chloro-4-iodobenzoic acid
Formel: C₇H₄ClIO₂
MW: 282,46 g/mol
MDL nummer: MFCD05863523
CAS-nummer: 145343-76-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 165.0 to 169.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR