(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride ≥96.0% (by total nitrogen and titration analysis)

Leverandør: TCI

Synonymer: (H-Cystine-OMe)₂·2HCl, L-Cystine dimethyl ester dihydrochloride, (H-Cys-OMe)₂·2 HCl, (H-Cys-OMe)2.2HCl

C3303-5G C3303-25G
TCIAC3303-5GEA 420 NOK
TCIAC3303-5G TCIAC3303-25G
(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride ≥96.0% (by total nitrogen and titration analysis)
(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride
Formel: C₈H₁₈Cl₂N₂O₄S₂
MW: 341,28 g/mol
MDL nummer: MFCD00012490
CAS-nummer: 32854-09-4
EINECS: 251-261-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01686194c34364a5ee3635d0d951765d2f200858&vid=d27273308284766b9978d8a5aa9957de950f3d8c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 96.0 %
Purity (Argentmetric Titration) min. 96.0 %
Specific rotation [a]20/D -39.0 to -43.0 deg (C=2,MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR