4-Chloro-2-fluoro-5-nitrotoluene ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
C3334-25G
C3334-5G
TCIAC3334-25GEA
4050
NOK
TCIAC3334-25G
TCIAC3334-5G
4-Chloro-2-fluoro-5-nitrotoluene ≥98.0% (by GC)
4-Chloro-2-fluoro-5-nitrotoluene
Formel:
C₇H₅ClFNO₂ MW: 189,57 g/mol |
MDL nummer:
MFCD00134231 CAS-nummer: 18349-11-6 EINECS: 242-225-9 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...