Ceftibuten hydrate ≥95.0% (by HPLC)

Leverandør: TCI

Synonymer: (6R,7R)-7-[[(2Z)-2-(2-Amino-4-thiazolyl)-4-carboxy-1-oxo-2-buten-1-yl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

C3391-100MG C3391-25MG
TCIAC3391-100MGEA 4350 NOK
TCIAC3391-100MG TCIAC3391-25MG
Ceftibuten hydrate ≥95.0% (by HPLC)
Ceftibuten hydrate
Formel: C₁₅H₁₄N₄O₆S₂·xH₂O
MW: 410,42 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00864918
CAS-nummer: 118081-37-1
Merck Indeks: 14,01949

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 95.0 area%
Specific rotation [a]20/D +135 to +155 deg (C=1, pH 8 Phosphate buffer (calcd.on anh.substance))
Water 8.0 to 13.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR