2-Cyanoisonicotinic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-Cyano-4-pyridinecarboxylic acid

C3396-200MG C3396-1G
TCIAC3396-200MGEA 825 NOK
TCIAC3396-200MG TCIAC3396-1G
2-Cyanoisonicotinic acid ≥98.0% (by GC, titration analysis)
2-Cyanoisonicotinic acid
Formel: C₇H₄N₂O₂
MW: 148,1200 g/mol
MDL nummer: MFCD03411729
CAS-nummer: 161233-97-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR