1-(2-Chlorophenyl)-3-methyl-5-pyrazolone ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

C3414-5G C3414-25G
TCIAC3414-5GEA 340 NOK
TCIAC3414-5G TCIAC3414-25G
1-(2-Chlorophenyl)-3-methyl-5-pyrazolone ≥98.0% (by HPLC, titration analysis)
1-(2-Chlorophenyl)-3-methyl-5-pyrazolone
Formel: C₁₀H₉ClN₂O
MW: 208,65 g/mol
MDL nummer: MFCD00059717
CAS-nummer: 14580-22-4
EINECS: 238-623-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1aa2c4ba83c0f137fea085547f0766401ea1094a&vid=2ef4b37c33e2e2c0a2c8a995f3a05e813176e843&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 198.0 to 202.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR