(3-Chloropropyl)tris(trimethylsilyloxy)silane ≥96.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-(3-Chloropropyl)-1,1,1,5,5,5-hexamethyl-3-[(trimethylsilyl)oxy]trisiloxane, (3-Chloropropyl)tris(trimethylsiloxy)silane, 3-(3-Chloropropyl)-1,1,1,5,5,5-hexamethyl-3-((trimethylsilyl)oxy)trisiloxane

C3437-1G C3437-5G
TCIAC3437-1GEA 220 NOK
TCIAC3437-1G TCIAC3437-5G
(3-Chloropropyl)tris(trimethylsilyloxy)silane ≥96.0% (by GC)
(3-Chloropropyl)tris(trimethylsilyloxy)silane
Formel: C₁₂H₃₃ClO₃Si₄
MW: 373,18 g/mol
Kokepunkt: 181°C
Flash punkt: 85°C
MDL nummer: MFCD00054781
CAS-nummer: 18077-31-1
EINECS: 241-985-9

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR