3-(Chlorosulphonyl)benzoic acid ≥96.0% (by titrimetric analysis)

Leverandør: TCI
Danger

C3466-25G C3466-5G
TCIAC3466-25GEA 1225 NOK
TCIAC3466-25G TCIAC3466-5G
3-(Chlorosulphonyl)benzoic acid ≥96.0% (by titrimetric analysis)
3-(Chlorosulphonyl)benzoic acid
Formel: C₇H₅ClO₄S
MW: 220,63 g/mol
MDL nummer: MFCD00024877
CAS-nummer: 4025-64-3
EINECS: 223-692-8
FN: 3261
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Argentmetric Titration) min. 96.0 %
Melting point 132.0 to 137.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR