4-Chloro-6-(4-chlorophenyl)-2-phenylpyrimidine ≥98.0% (by GC)

Leverandør: TCI
Warning

C3518-200MG C3518-1G
TCIAC3518-200MGEA 460 NOK
TCIAC3518-200MG TCIAC3518-1G
4-Chloro-6-(4-chlorophenyl)-2-phenylpyrimidine ≥98.0% (by GC)
4-Chloro-6-(4-chlorophenyl)-2-phenylpyrimidine
Formel: C₁₆H₁₀Cl₂N₂
MW: 301,17 g/mol
MDL nummer: MFCD30536778
CAS-nummer: 1354749-13-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 129.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR