4-Carboxyphenylacetic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: (4-Carboxyphenyl)acetic acid, 4-(Carboxymethyl)benzoic acid

C3530-5G C3530-25G
TCIAC3530-5GEA 595 NOK
TCIAC3530-5G TCIAC3530-25G
4-Carboxyphenylacetic acid ≥98.0% (by titrimetric analysis)
4-Carboxyphenylacetic acid
Formel: C₉H₈O₄
MW: 180,16 g/mol
MDL nummer: MFCD01074834
CAS-nummer: 501-89-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 243.0 to 247.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR