4-Cyano-3-fluorophenyl 4-Propylbenzoate ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4-Cyano-3-fluorophenyl 4-propylbenzoate, 4-Propylbenzoic acid 4-Cyano-3-fluorophenyl ester

C3554-1G C3554-5G
TCIAC3554-1GEA 325 NOK
TCIAC3554-1G TCIAC3554-5G
4-Cyano-3-fluorophenyl 4-Propylbenzoate ≥98.0% (by GC)
4-Cyano-3-fluorophenyl 4-Propylbenzoate
Formel: C₁₇H₁₄FNO₂
MW: 283.30 g/mol
MDL nummer: MFCD06797522
CAS-nummer: 86776-51-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR