SCR7 pyrazine ≥98.0% (by HPLC)
Leverandør: TCI
Synonymer:
2,3-Dihydro-6,7-diphenyl-2-thioxo-4(1H)-pteridinone, 6,7-Diphenyl-2-thioxo-2,3-dihydropteridin-4(1H)-one
C3577-10MG
C3577-50MG
TCIAC3577-10MGEA
1425
NOK
TCIAC3577-10MG
TCIAC3577-50MG
SCR7 pyrazine ≥98.0% (by HPLC)
SCR7 pyrazine
Formel:
C₁₈H₁₂N₄OS MW: 332,38 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD02167478 CAS-nummer: 14892-97-8 |
Spesifikasjon Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 207.0 to 211.0 deg-C |
Elemental analysis (Nitrogen) | 16.00 to 17.50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...