Creatine (N-amidinosarcosine) ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 2-(1-Methylguanidino)acetic acid, N-Amidinosarcosine, Creatine

C3610-100G C3610-25G
TCIAC3610-100GEA 1100 NOK
TCIAC3610-100G TCIAC3610-25G
Creatine (N-amidinosarcosine) ≥98.0% (by HPLC, titration analysis)
Creatine (N-amidinosarcosine)
Formel: C₄H₉N₃O₂
MW: 131,13 g/mol
Smeltepunkt: 303°C
MDL nummer: MFCD00004282
CAS-nummer: 57-00-1
EINECS: 200-306-6
Merck Indeks: 14,02568

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR