(3-Chloro-4-methoxyphenyl)methanamine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
3-Chloro-4-methoxybenzylamine
C3634-1G
C3634-5G
TCIAC3634-1GEA
350
NOK
TCIAC3634-1G
TCIAC3634-5G
(3-Chloro-4-methoxyphenyl)methanamine ≥98.0% (by GC, titration analysis)
(3-Chloro-4-methoxyphenyl)methanamine
Formel:
C₈H₁₀ClNO MW: 171,63 g/mol Kokepunkt: 110°C |
MDL nummer:
MFCD00995395 CAS-nummer: 115514-77-7 FN: 2735 ADR: 8,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...