4-(3-Chloro-4-fluorophenylamino)-7-methoxyquinazolin-6-ol ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: FAAH-IN-2, O-Desmorpholinopropyl Gefitinib, 4-(3-Chloro-4-fluoroanilino)-6-hydroxy-7-methoxyquinazoline, 4-((3-Chloro-4-fluorophenyl)amino)-7-methoxyquinazolin-6-ol

C3647-1G C3647-5G
TCIAC3647-1GEA 380 NOK
TCIAC3647-1G TCIAC3647-5G
4-(3-Chloro-4-fluorophenylamino)-7-methoxyquinazolin-6-ol ≥98.0% (by HPLC, titration analysis)
4-(3-Chloro-4-fluorophenylamino)-7-methoxyquinazolin-6-ol
Formel: C₁₅H₁₁ClFN₃O₂
MW: 319,72 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD09908046
CAS-nummer: 184475-71-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR