1,4-Cineole ≥93.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1-Isopropyl-4-methyl-7-oxabicyclo[2.2.1]heptane

C3652-100G C3652-25G
TCIAC3652-100GEA 1475 NOK
TCIAC3652-100G TCIAC3652-25G
1,4-Cineole ≥93.0% (by GC)
1,4-Cineole
Formel: C₁₀H₁₈O
MW: 154,25 g/mol
Kokepunkt: 173°C
Smeltepunkt: –46°C
Flash punkt: 47°C
MDL nummer: MFCD00209502
CAS-nummer: 470-67-7
EINECS: 207-428-9
FN: 1993
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR