2-[2-(Diethylamino)ethoxy]ethanol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Ethylene Glycol Mono[2-(diethylamino)ethyl] Ether, 2-(2-(Diethylamino)ethoxy)ethanol

D0298-25G D0298-500G
TCIAD0298-25GEA 365 NOK
TCIAD0298-25G TCIAD0298-500G
2-[2-(Diethylamino)ethoxy]ethanol ≥98.0% (by GC)
2-[2-(Diethylamino)ethoxy]ethanol
Formel: C₈H₁₉NO₂
MW: 161,25 g/mol
Kokepunkt: 101°C
MDL nummer: MFCD00020604
CAS-nummer: 140-82-9
EINECS: 205-436-7

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR