3,5-Diiodo-L-tyrosine dihydrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: H-Tyr(3,5-DiI)-OH·2H₂O, (S)-2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid dihydrate

D0612-25G D0612-5G
TCIAD0612-25GEA 945 NOK
TCIAD0612-25G TCIAD0612-5G
3,5-Diiodo-L-tyrosine dihydrate ≥98.0% (by titrimetric analysis)
3,5-Diiodo-L-tyrosine dihydrate
Formel: C₉H₉I₂NO₃·2H₂O
MW: 469,01 g/mol
MDL nummer: MFCD00016542
CAS-nummer: 18835-59-1
EINECS: 206-092-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea2758371e6ad1e96805ddd1c42ce6438dac37c5&vid=bea71cf8f54f1ff713a3ec5628068648e924282d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (as Dihydrate)
Specific rotation [a]20/D +0.8 to +1.5 deg (C=5, 1mol/L HCl)
Solubility in NaOH (100g/L) very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR