2,4-Diamino-6-diallylamino-1,3,5-triazine ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
N2,N2-Diallylmelamine
D1075-25G
D1075-500G
TCIAD1075-25GEA
570
NOK
TCIAD1075-25G
TCIAD1075-500G
2,4-Diamino-6-diallylamino-1,3,5-triazine ≥98.0% (by HPLC, titration analysis)
2,4-Diamino-6-diallylamino-1,3,5-triazine
Formel:
C₉H₁₄N₆ MW: 206,25 g/mol |
MDL nummer:
MFCD00047347 CAS-nummer: 91-77-0 EINECS: 202-096-1 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Bromination Method) | min. 98.0 % |
Melting point | 143.0 to 146.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...