Bis(3-pyridylmethyl)amine ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: alpha,alpha'-Iminodi(3-picoline), 3,3'-Dipicolylamine

D1199-25ML D1199-5ML
TCIAD1199-25MLEA 6075 NOK
TCIAD1199-25ML TCIAD1199-5ML
Bis(3-pyridylmethyl)amine ≥97.0% (by GC)
Bis(3-pyridylmethyl)amine
Formel: C₁₂H₁₃N₃
MW: 199,26 g/mol
MDL nummer: MFCD00038043
CAS-nummer: 1656-94-6
EINECS: 216-753-5

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR