Diphenolic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4,4-Bis(4-hydroxyphenyl)valeric Acid
D1274-25G
D1274-500G
TCIAD1274-25GEA
380
NOK
TCIAD1274-25G
TCIAD1274-500G
Diphenolic acid ≥98.0% (by GC, titration analysis)
Diphenolic acid
Formel:
C₁₇H₁₈O₄ MW: 286,33 g/mol |
MDL nummer:
MFCD00002800 CAS-nummer: 126-00-1 EINECS: 204-763-2 Merck Indeks: 14,03312 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 174.0 to 177.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...