1,3-Dipropoxybenzene ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Resorcinol Dipropyl Ether
D1362-1ML
D1362-5ML
TCIAD1362-1MLEA
700
NOK
TCIAD1362-1ML
TCIAD1362-5ML
1,3-Dipropoxybenzene ≥98.0% (by GC)
1,3-Dipropoxybenzene
Formel:
C₁₂H₁₈O₂ MW: 194,27 g/mol Kokepunkt: 251°C |
MDL nummer:
MFCD00059402 CAS-nummer: 56106-37-7 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...