Dansyl-L-leucine ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Synonymer:
5-Dimethylaminonaphthalene-1-sulphonyl-L-leucine, Dns-Leu-OH
D1498-1G
D1498-100MG
TCIAD1498-1GEA
2200
NOK
TCIAD1498-1G
TCIAD1498-100MG
Dansyl-L-leucine ≥98.0% (by titrimetric analysis)
Dansyl-L-leucine
Formel:
C₁₈H₂₄N₂O₄S MW: 364,46 g/mol |
MDL nummer:
MFCD00069532 CAS-nummer: 1100-22-7 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 124.0 to 127.0 deg-C |
Specific rotation [a]20/D | -8.0 to -11.0 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...