S,S'-Dimethyl-N-cyanodithioiminocarbonate ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Danger

Synonymer: Dimethyl N-Cyanodithioiminocarbonate, Dimethyl N-Cyanodithiocarbonimidate

D1798-25G D1798-250G
TCIAD1798-25GEA 420 NOK
TCIAD1798-25G TCIAD1798-250G
S,S'-Dimethyl-N-cyanodithioiminocarbonate ≥98.0% (by total nitrogen basis)
S,S'-Dimethyl-N-cyanodithioiminocarbonate
Formel: C₄H₆N₂S₂
MW: 146,23 g/mol
MDL nummer: MFCD00009825
CAS-nummer: 10191-60-3
EINECS: 233-467-6
FN: 1759
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Purity (with Total Nitrogen) min. 98.0 %
Melting point 53.0 to 58.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR