Pentasodium diethylenetriaminepentaacetate ∼40% ∼1.0 mol/L in water

Leverandør: TCI

Synonymer: Diethylenetriaminepentaacetic Acid Pentasodium Salt

D1870-25G D1870-500G
TCIAD1870-25GEA 220 NOK
TCIAD1870-25G TCIAD1870-500G
Pentasodium diethylenetriaminepentaacetate ∼40% ∼1.0 mol/L in water
Pentasodium diethylenetriaminepentaacetate
Formel: C₁₄H₂₃N₃O₁₀
MW: 503,26 g/mol
MDL nummer: MFCD00051016
CAS-nummer: 140-01-2
EINECS: 205-391-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=145eff48412750378428a6ac86ccc7a72253ea69&vid=06206f7accf858111ecc285338f356bf0b0187c3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green clear liquid
Concentration (Chelometric Method)unit2 38.0 to 41.8 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR