Diphenyl (2,3-dihydro-2-thioxo-3-benzoxazolyl)phosphonate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: (2,3-Dihydro-2-thioxo-3-benzoxazolyl)phosphonic Acid Diphenyl Ester, DBOP

D2038-25G D2038-5G
TCIAD2038-25GEA 3725 NOK
TCIAD2038-25G TCIAD2038-5G
Diphenyl (2,3-dihydro-2-thioxo-3-benzoxazolyl)phosphonate ≥98.0% (by titrimetric analysis)
Diphenyl (2,3-dihydro-2-thioxo-3-benzoxazolyl)phosphonate
Formel: C₁₉H₁₄NO₄PS
MW: 383,36 g/mol
MDL nummer: MFCD00142793
CAS-nummer: 111160-56-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 81.0 to 86.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR